Learn More
ABclonal Technology MCT4/SLC16A3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A105481000UL
Encompass_Preferred
Lactic acid and pyruvate transport across plasma membranes is catalyzed by members of the proton-linked monocarboxylate transporter (MCT) family, which has been designated solute carrier family-16. Each MCT appears to have slightly different substrate and inhibitor specificities and transport kinetics, which are related to the metabolic requirements of the tissues in which it is found. The MCTs, which include MCT1 (SLC16A1, MIM 600682) and MCT2 (SLC16A7, MIM 603654), are characterized by 12 predicted transmembrane domains (Price et al., 1998 [PubMed 9425115]).
Catalog No. 50-001-35794