Learn More
ABclonal Technology IL17C Rabbit pAb
SDP
Supplier:  ABclonal Technology A105871000UL
Encompass_Preferred
The protein encoded by this gene is a T cell-derived cytokine that shares the sequence similarity with IL17. This cytokine was reported to stimulate the release of tumor necrosis factor alpha and interleukin 1 beta from a monocytic cell line. The expression of this cytokine was found to be restricted to activated T cells.
Catalog No. 50-001-38290