Learn More
ABclonal Technology TMPRSS6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1064220UL
Encompass_Preferred
The protein encoded by this gene is a type II transmembrane serine proteinase that is found attached to the cell surface. The encoded protein may be involved in matrix remodeling processes in the liver. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-37855