Learn More
ABclonal Technology UCK2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10815500UL
Encompass_Preferred
This gene encodes a pyrimidine ribonucleoside kinase. The encoded protein (EC 2.7.1.48) catalyzes phosphorylation of uridine and cytidine to uridine monophosphate (UMP) and cytidine monophosphate (CMP), respectively.
Catalog No. 50-001-37814