Learn More
Cayman Chemical A11trIfluoroacetate salt 5mg
SDP
Supplier:  Cayman Chemical 356535 MG
Encompass_Preferred
A cell-penetrating peptide; decreases annexin A1 binding to EphA2 and increases EphA2 ubiquitination in HK1 NPC cells; inhibits HK1 and 5-8F NPC cell proliferation, migration, and invasion; reduces tumor volume in 5-8F and HK1 NPC mouse xenograft models
Catalog No. 50-291-0179