Learn More
ABclonal Technology SLC18A3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A160681000UL
Encompass_Preferred
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Catalog No. 50-001-12582