Learn More
ABclonal Technology DNTTIP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161201000UL
Encompass_Preferred
This gene is thought to be involved in chromatin remodeling and gene transcription. The encoded nuclear protein binds to and enhances the transcriptional activity of the estrogen receptor alpha, and also interacts with terminal deoxynucleotidyltransferase. The expression profile of this gene is a potential biomarker for chronic obstructive pulmonary disease.
Catalog No. 50-001-11713