Learn More
ABclonal Technology CAB39 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161251000UL
Encompass_Preferred
Enables kinase binding activity and protein serine/threonine kinase activator activity. Involved in intracellular signal transduction, peptidyl-serine phosphorylation, and positive regulation of protein phosphorylation. Located in extracellular exosome. Implicated in hepatocellular carcinoma. Biomarker of hepatocellular carcinoma and pancreatic cancer.
Catalog No. 50-001-11900