Learn More
ABclonal Technology IL23R Rabbit pAb
SDP
Supplier:  ABclonal Technology A1613500UL
Encompass_Preferred
The protein encoded by this gene is a subunit of the receptor for IL23A/IL23. This protein pairs with the receptor molecule IL12RB1/IL12Rbeta1, and both are required for IL23A signaling. This protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner.
Catalog No. 50-001-11693