Learn More
ABclonal Technology TAAR1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16166500UL
Encompass_Preferred
The protein encoded by this gene is a G-protein coupled receptor activated by trace amines. The encoded protein responds little or not at all to dopamine, serotonin, epinephrine, or histamine, but responds well to beta-phenylethylamine, p-tyramine, octopamine, and tryptamine. While primarily functioning in neurologic systems, there is evidence that this gene is involved in blood cell and immunologic functions as well. This gene is thought to be intronless.
Catalog No. 50-001-11666