Learn More
ABclonal Technology HCN1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161981000UL
Encompass_Preferred
The membrane protein encoded by this gene is a hyperpolarization-activated cation channel that contributes to the native pacemaker currents in heart and neurons. The encoded protein can homodimerize or heterodimerize with other pore-forming subunits to form a potassium channel. This channel may act as a receptor for sour tastes.
Catalog No. 50-001-10469