Learn More
ABclonal Technology KIAA1429 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16201500UL
Encompass_Preferred
Enables RNA binding activity. Involved in mRNA alternative polyadenylation and mRNA methylation. Located in cytosol and nuclear speck. Colocalizes with RNA N6-methyladenosine methyltransferase complex.
Catalog No. 50-001-10104