Learn More
ABclonal Technology CCL7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A162211000UL
Encompass_Preferred
This gene encodes monocyte chemotactic protein 3, a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix. This gene is part of a cluster of C-C chemokine family members on chromosome 17q.
Catalog No. 50-001-12934