Learn More
ABclonal Technology [KO Validated] KMT5C Rabbit pAb
SDP
Supplier:  ABclonal Technology A162351000UL
Encompass_Preferred
SUV420H2 and the related enzyme SUV420H1 (MIM 610881) function as histone methyltransferases that specifically trimethylate nucleosomal histone H4 (see MIM 602822) on lysine-20 (K20) (Schotta et al., 2004 [PubMed 15145825]).
Catalog No. 50-001-12536