Learn More
ABclonal Technology ACSL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A162531000UL
Encompass_Preferred
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. Several transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-11611