Learn More
ABclonal Technology VDAC2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16294500UL
Encompass_Preferred
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
Catalog No. 50-001-11662