Learn More
ABclonal Technology KLF8 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16321500UL
Encompass_Preferred
This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-09931