Learn More
ABclonal Technology SUPT6H Rabbit pAb
SDP
Supplier:  ABclonal Technology A164341000UL
Encompass_Preferred
Enables histone binding activity. Involved in negative regulation of histone H3-K27 methylation and positive regulation of transcription elongation from RNA polymerase II promoter. Predicted to be located in nucleoplasm. Predicted to be part of transcription elongation factor complex. Predicted to be active in transcriptionally active chromatin.
Catalog No. 50-001-11017