Learn More
ABclonal Technology ZNF263 Rabbit pAb
SDP
Supplier:  ABclonal Technology A164771000UL
Encompass_Preferred
Enables DNA-binding transcription repressor activity, RNA polymerase II-specific and transcription cis-regulatory region binding activity. Involved in negative regulation of transcription by RNA polymerase II and positive regulation of transcription by RNA polymerase II. Located in nucleus.
Catalog No. 50-001-11945