Learn More
ABclonal Technology KLF2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16480500UL
Encompass_Preferred
This gene encodes a protein that belongs to the Kruppel family of transcription factors. The encoded zinc finger protein is expressed early in mammalian development and is found in many different cell types. The protein acts to bind the CACCC box found in the promoter of target genes to activate their transcription. It plays a role in many processes during development and disease including adipogenesis, embryonic erythropoiesis, epithelial integrity, inflammation and t-cell viability.
Catalog No. 50-001-09435