Learn More
ABclonal Technology SRSF1/SF2/ASF Rabbit pAb
SDP
Supplier:  ABclonal Technology A1649500UL
Encompass_Preferred
This gene encodes a member of the arginine/serine-rich splicing factor protein family. The encoded protein can either activate or repress splicing, depending on its phosphorylation state and its interaction partners. Multiple transcript variants have been found for this gene. There is a pseudogene of this gene on chromosome 13.
Catalog No. 50-001-10417