Learn More
ABclonal Technology CTNNA3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A165191000UL
Encompass_Preferred
This gene encodes a protein that belongs to the vinculin/alpha-catenin family. The encoded protein plays a role in cell-cell adhesion in muscle cells. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia, familial 13. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-12861