Learn More
ABclonal Technology UGT2B15 Rabbit pAb
SDP
Supplier:  ABclonal Technology A166571000UL
Encompass_Preferred
This gene encodes a glycosyltransferase that is invovled in the metabolism and elimination of toxic compounts, both endogenous and of xenobiotic origin. This gene plays a role in the regulation of estrogens and androgens. This locus is present in a cluster of similar genes and pseudogenes on chromosome 4.
Catalog No. 50-001-10583