Learn More
ABclonal Technology CAMKK1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16716500UL
Encompass_Preferred
The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This protein plays a role in the calcium/calmodulin-dependent (CaM) kinase cascade. Three transcript variants encoding two distinct isoforms have been identified for this gene.
Catalog No. 50-001-12740