Learn More
ABclonal Technology MAPK1/MAPK3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16736500UL
Encompass_Preferred
MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
Catalog No. 50-001-09564