Learn More
ABclonal Technology CCKAR Rabbit pAb
SDP
Supplier:  ABclonal Technology A167991000UL
Encompass_Preferred
This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin (CCK) family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
Catalog No. 50-001-10711