Learn More
Medchemexpress LLC A17 | 2568186-47-8 | 99.4% | 533.75 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY163649
Encompass_Preferred
A17 is a bile acid analog with anti-non-alcoholic steatohepatitis (NASH) and anti-inflammatory activities. It reduces fatty acid (FA) uptake and promotes FA oxidation by inhibiting fatty acid translocase (Cd36) expression and activating AMPKα. A17 can be used for NASH research.
  • Bile acid analog
  • Anti-non-alcoholic steatohepatitis (NASH) activity
  • Anti-inflammatory activity
  • Reduces fatty acid (FA) uptake
  • Promotes fatty acid oxidation
  • Inhibits fatty acid translocase (Cd36) expression
  • Activates AMPKα
  • Suitable for NASH research
Catalog No. 50-003-92869