Learn More
Aviva Systems Biology Corp A1BG Antibody - N-terminal region (ARP33810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A1BG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human A1BG. Alternate names: A1B, ABG, GAB, HYST2477, A1BGApplicationWB. Species reactivity: Human
Catalog No. 50-014-55156