Learn More
QA Bio Inc Ludger A2 Glycan - 20µg
Supplier:  QA Bio Inc CNA220U
Encompass
A2 Glycan Synonyms: A2 N-linked oligosaccharide. G2S2, A2G2S2, A2G(4)2S(6)2. • Description: Di-sialylated, bi-antennary complex-type N-glycan (oligosaccharide). • Molecular Weight: 2224 • Purity: >90% pure as assessed by a combination of 1 H-NMR and HPLC. • Sources: A2 glycan is found on many mammalian glycoproteins including serum transferrin, and fibrin. This product is typically purified from the oligosaccharide pool released from fibrin by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. Contains ammonium salt to stabilise against desialylation.
Catalog No. 50-223-6587