Learn More
ATCC A20; B Cell Lymphoma; Mouse (Mus musculus)
Supplier:  ATCC TIB208
Encompass
A20 [A-20] is a BALB/c B cell lymphoma line derived from a spontaneous reticulum cell neoplasm found in an old BALB/cAnN mouse. This cell line can be used in immunology research.
Catalog No. 50-238-3975