Learn More
ABclonal Technology Phospho-SQSTM1/p62-S349 Rabbit pAb
SDP
Supplier:  ABclonal Technology A207371000UL
Encompass_Preferred
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B (NF-kB) signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Catalog No. 50-001-13330