Learn More
Alkali Scientific A23187 [Calcium Ionophore][Calcimycin], 25 Milligrams
SDP
Supplier:  Alkali Scientific ACA002CP
Encompass_Preferred
Calcium Ionophore A23187, also known as calcimycin, is a mobile ion carrier used in research to transport calcium ions across biological membranes. It facilitates the movement of calcium into cells, thereby increasing intracellular calcium levels and enabling the study of calcium dependent cellular processes. This compound is widely used in cell biology and biochemical research to investigate signal transduction, enzyme activation, and cellular responses regulated by calcium signaling pathways. It is particularly valuable in experiments requiring controlled manipulation of calcium flux. The reagent is commonly applied in pharmacological studies and advanced research workflows focusing on cellular physiology and signaling mechanisms.
Catalog No. 50-006-60417