Learn More
Alkali Scientific A23187 [Calcium Ionophore][Calcimycin], 5 Milligrams
SDP
Supplier:  Alkali Scientific ACA001CP
Encompass_Preferred
Calcimycin (A23187) is a calcium ionophore widely used in biochemical and cell signaling research to facilitate the transport of calcium ions across biological membranes. It increases intracellular calcium levels by forming complexes with calcium ions and enabling their movement through lipid membranes. This property makes it a valuable tool for studying calcium dependent cellular processes such as signal transduction, enzyme activation, and secretion pathways. Calcimycin is commonly used in pharmacological research, cell biology experiments, and studies involving calcium regulation. It helps researchers investigate cellular responses triggered by calcium flux and is frequently applied in controlled laboratory settings for mechanistic studies.
Catalog No. 50-006-60400