Learn More
Medchemexpress LLC A2AR-agonist-1 | 41552-95-8 | 99.9% | 410.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY187761MG
Encompass_Preferred
A2AR-agonist-1 is a potent A2AR agonist and ENT1 inhibitor with Ki of 4.39 and 3.47 for A2AR and ENT1. It is for research use only.
  • Potent A2AR agonist
  • ENT1 inhibitor
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-88251