Learn More
QA Bio Inc Ludger A2F Glycan - 20µg
Supplier:  QA Bio Inc CNA2F20U
Encompass
A2F Glycan Synonyms: A2 N-linked oligosaccharide. G2FS2, FA2G2S2, F(6)A2G(4)2S(6)2. • Description: Di-sialylated, core-fucosylated bi-antennary complex-type N-glycan (oligosaccharide). • Molecular Weight: 2370 • Purity: >90% pure as assessed by a combination of 1 H-NMR and HPLC. • Sources: A2F glycan is found on many mammalian glycoproteins including thyroglobulin, gamma globulins, and IgG. This product is typically purified from the oligosaccharide pool released from porcine thyroglobulin by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. Contains ammonium salt to stabilise against desialylation.
Catalog No. 50-223-6588