Learn More
Biolegend Alexa Fluor 488 anti-human CD103 (Integrin αE)
Supplier:  Biolegend 350208
Encompass_Preferred
Alexa Fluor 488 anti-human CD103 (Integrin αE) [Ber-ACT8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-168-584