Learn More
Biolegend Alexa Fluor 488 anti-human CD3
Supplier:  Biolegend 344810
Encompass_Preferred
Alexa Fluor 488 anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-168-305