Learn More
Biolegend Alexa Fluor 488 anti-mouse CD117 (c-kit)
Supplier:  Biolegend 105816
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. NC0155875