Learn More
Biolegend Alexa Fluor 488 anti-mouse CD4
Supplier:  Biolegend 100425
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, ICC; Size: 25 ug
Catalog No. 50-162-427