Learn More
Biolegend Alexa Fluor 488 anti-mouse F4/80
Supplier:  Biolegend 123119
Encompass_Preferred
Alexa Fluor 488 anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-164-064