Learn More
ABclonal Technology JunD Rabbit mAb
SDP
Supplier:  ABclonal Technology A5433500UL
Encompass_Preferred
The protein encoded by this intronless gene is a member of the JUN family, and a functional component of the AP1 transcription factor complex. This protein has been proposed to protect cells from p53-dependent senescence and apoptosis. Alternative translation initiation site usage results in the production of different isoforms (PMID12105216).
Catalog No. 50-001-36264