Learn More
ABclonal Technology CRABP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A54341000UL
Encompass_Preferred
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
Catalog No. 50-001-37015