Learn More
ATCC A549 Exosomes; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL185EXM
Encompass
Exosomes are subcellular nanoparticles of size 50-200 nm released from cells through a fusion of multicellular bodies with plasma membrane. The exosomes are purified from cell culture medium of lung carcinoma cell line A549 (ATCC CCL-185). The exosomes are purified and concentrated using tangential flow filtration system in a sterile environment. The purified exosomes were further filtered through 0.2 micron sterile filters.
Catalog No. 50-238-2089