Learn More
Cell Biolabs Inc A549/GFP Cell Line
Supplier:  Cell Biolabs Inc AKR209
Encompass_Preferred
Cell Biolabs' A549/GFP Cell Line stably expresses GFP which was introduced into parental A549 lung cancer cells using lentivirus. This cell line otherwise exhibits the same characteristics of the parental A549 cell line.
Catalog No. 50-672-592