Learn More
ATCC A-549; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL185
Encompass
A549 cells were isolated from the lung tissue of a White, 58-year-old male with lung cancer. This cell line can be used in cancer, immuno-oncology, and toxicology research.
Catalog No. 50-238-4533
 Restricted Item