Learn More
ABclonal Technology HSPE1/HSP10/CPN10 Rabbit mAb
SDP
Supplier:  ABclonal Technology A5580500UL
Encompass_Preferred
This gene encodes a major heat shock protein which functions as a chaperonin. Its structure consists of a heptameric ring which binds to another heat shock protein in order to form a symmetric, functional heterodimer which enhances protein folding in an ATP-dependent manner. This gene and its co-chaperonin, HSPD1, are arranged in a head-to-head orientation on chromosome 2. Naturally occurring read-through transcription occurs between this locus and the neighboring locus MOBKL3.
Catalog No. 50-001-37016