Learn More
ABclonal Technology NME2/NM23B Rabbit mAb
SDP
Supplier:  ABclonal Technology A5621500UL
Encompass_Preferred
Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of A (encoded by NME1) and B (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants have been found for this gene. Read-through transcription from the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) that encode a fusion protein comprised of sequence sharing identity with each individual gene product.
Catalog No. 50-001-36307