Learn More
ABclonal Technology CEND1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A5930500UL
Encompass_Preferred
The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.
Catalog No. 50-001-35536