Learn More
ABclonal Technology PERP Rabbit mAb
SDP
Supplier:  ABclonal Technology A5937500UL
Encompass_Preferred
Involved in activation of cysteine-type endopeptidase activity. Predicted to be located in plasma membrane. Predicted to be active in cell-cell junction. Implicated in erythrokeratodermia variabilis and mutilating palmoplantar keratoderma with periorificial keratotic plaques.
Catalog No. 50-001-34576