Learn More
Biolegend Alexa Fluor 594 anti-mouse CD4
Supplier:  Biolegend 100446
Encompass_Preferred
Alexa Fluor 594 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: IHC-F, FC, ICC; Size: 100 ug
Catalog No. 50-711-802